Gene Transcripts

Gene Symbol
Phox2b
Entrez Gene ID
18935
Gene Name
paired-like homeobox 2b
Gene Aliases
Dilp1, NBPhox, Pmx2b
Chromosome Location
Chr. 5: 67094397 - 67099126 on Build GRCm38
UniGene ID
Mm.62505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008888.3 NP_032914.1 3 1421
GenBank mRNA BC079610.1 - 3 1421
Y14493.1 - 3 1453

View Full Product Details