Gene Transcripts

Gene Symbol
Mapk10
Entrez Gene ID
26414
Gene Name
mitogen-activated protein kinase 10
Gene Aliases
C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK
Chromosome Location
Chr. 5: 102907948 - 103211339 on Build GRCm38
UniGene ID
Mm.39253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081567.1 NP_001075036.1 3, 4 640
NM_009158.2 NP_033184.2 3, 4 640
GenBank mRNA AB096076.1 - 1, 2 74
AB096077.1 - 1, 2 74
AB096078.1 - 1, 2 74
AB096079.1 - 1, 2 74
AK042368.1 - 2, 3 205
AK082242.1 - 3, 4 643
AK163206.1 - 3, 4 591
BC046625.1 - 3, 4 579 6494163
L35236.1 - 3, 4 383

View Full Product Details