Gene Transcripts

Gene Symbol
Slc16a2
Entrez Gene ID
20502
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 2
Gene Aliases
AW105741, Mct8, Xpct
Chromosome Location
Chr. X: 103697414 - 103821988 on Build GRCm38
UniGene ID
Mm.388973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009197.2 NP_033223.2 1 605
GenBank mRNA AB265788.1 - 1 431
AF045692.1 - 1 653
BC080678.1 - 1 441 6313495
BC096587.1 - 1 541 3992886
BM935188.1 - 1 118
CX218841.1 - 1 541

View Full Product Details