Gene Transcripts

Gene Symbol
Tpst2
Entrez Gene ID
22022
Gene Name
protein-tyrosine sulfotransferase 2
Gene Aliases
AI448750, D5Ucla3, Tango13b, grm, grt
Chromosome Location
Chr. 5: 112276682 - 112315356 on Build GRCm38
UniGene ID
Mm.250631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009419.3 NP_033445.2 5, 6 1296
GenBank mRNA AF049890.1 - 4, 5 1242
AK036436.1 - 4, 5 1225
AK075672.1 - 5, 6 1296
AK149502.1 - 5, 6 1261
AK150082.1 - 5, 6 1256
AK163461.1 - 5, 6 1260
AK204832.1 - 1, 2 51
BC003721.1 - 5, 6 1287 3494892

View Full Product Details