Gene Transcripts

Gene Symbol
Crisp2
Entrez Gene ID
22024
Gene Name
cysteine-rich secretory protein 2
Gene Aliases
CRISP-2, GAPDL5, Tpx-1, Tpx1
Chromosome Location
Chr. 17: 40764734 - 40794146 on Build GRCm38
UniGene ID
Mm.1296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204071.1 NP_001191000.1 10 950
NM_009420.2 NP_033446.1 10 968
GenBank mRNA BC049615.1 - 10 920
M25533.1 - 10 920

View Full Product Details