Gene Transcripts

Gene Symbol
Neurod6
Entrez Gene ID
11922
Gene Name
neurogenic differentiation 6
Gene Aliases
Atoh2, Math-2, Math2, Nex, Nex1m, bHLHa2
Chromosome Location
Chr. 6: 55677818 - 55681263 on Build GRCm38
UniGene ID
Mm.5106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009717.2 NP_033847.1 2 407
GenBank mRNA AK144303.1 - 2 191
AK144360.1 - 2 408
AK144377.1 - 2 242
BC087831.1 - 2 200 6411826
U29086.1 - 2 213

View Full Product Details