Gene Transcripts

Gene Symbol
Csf2
Entrez Gene ID
12981
Gene Name
colony stimulating factor 2 (granulocyte-macrophage)
Gene Aliases
Csfgm, GMCSF, Gm-CSf, MGI-IGM
Chromosome Location
Chr. 11: 54247270 - 54249899 on Build GRCm38
UniGene ID
Mm.4922
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC116878.1 - 4 524
BC116880.1 - 4 524 40126199
X02333.1 - 4 674
X03019.1 - 4 533

View Full Product Details