Gene Transcripts

Gene Symbol
S1pr4
Entrez Gene ID
13611
Gene Name
sphingosine-1-phosphate receptor 4
Gene Aliases
Edg6, Lpc1, S1p4
Chromosome Location
Chr. 10: 81497745 - 81500137 on Build GRCm38
UniGene ID
Mm.33065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010102.2 NP_034232.1 1 1119
GenBank mRNA AJ006074.1 - 1 1119
AK075854.1 - 1 638
AK154242.1 - 1 642
BC107000.1 - 1 689 40045304

View Full Product Details