Gene Transcripts
- Gene Symbol
- Ifna9
- Entrez Gene ID
- 15972
- Gene Name
- interferon alpha 9
- Gene Aliases
- Ifa9
- Chromosome Location
- Chr. 4: 88591813 - 88592385 on Build GRCm38
- UniGene ID
- Mm.377092
- Species
- Mus musculus
- Gene Symbol
- Ifna6
- Entrez Gene ID
- 15969
- Gene Name
- interferon alpha 6
- Gene Aliases
- IFNa8/6, Ifa6, Ifa8, Ifna8
- Chromosome Location
- Chr. 4: 88827416 - 88827985 on Build GRCm38
- UniGene ID
- Mm.377090
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC117774.1 |
- |
|
106 |
|
View Full Product Details