Gene Transcripts

Gene Symbol
Mtl5
Entrez Gene ID
17771
Gene Name
metallothionein-like 5, testis-specific (tesmin)
Gene Aliases
tesmin
Chromosome Location
Chr. 19: 3388857 - 3407823 on Build GRCm38
UniGene ID
Mm.209941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039657.2 NP_001034746.1 6, 7 1022
NM_001286557.1 NP_001273486.1 7, 8 1106
NM_001286558.1 NP_001273487.1 6, 7 1037
GenBank mRNA AB057423.1 - 6, 7 1010
AF329359.1 - 6, 7 1047
AF329360.1 - 7, 8 1116
AK015724.1 - 6, 7 962
AK161511.1 - 6, 7 1023
BC024377.1 - 6, 7 998 4976492
U67176.1 - 8, 9 1039
U77383.1 - 6, 7 659

View Full Product Details