Gene Transcripts

Gene Symbol
Pa2g4
Entrez Gene ID
18813
Gene Name
proliferation-associated 2G4
Gene Aliases
38kDa, AA672939, Ebp1, Plfap
Chromosome Location
Chr. 10: 128557766 - 128565934 on Build GRCm38
UniGene ID
Mm.4742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011119.3 NP_035249.1 2, 3 319
GenBank mRNA AK002685.1 - 2, 3 397
AK076286.1 - 2, 3 324
AK144645.1 - 2, 3 373
AK166936.1 - 2, 3 328
AK168582.1 - 2, 3 321
BC003777.1 - 2, 3 309
BC005593.1 - 2, 3 309
BC028926.1 - 2, 3 302
BC037623.1 - 2, 3 339
BC046532.1 - 2, 3 273 5358060
U43918.1 - 2, 3 478
X84789.1 - 2, 3 384

View Full Product Details