Gene Transcripts

Gene Symbol
Pou2f1
Entrez Gene ID
18986
Gene Name
POU domain, class 2, transcription factor 1
Gene Aliases
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1
Chromosome Location
Chr. 1: 165865150 - 166002672 on Build GRCm38
UniGene ID
Mm.245261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011137.3 NP_035267.2 4 275
NM_198932.3 NP_945150.2 4 275
NM_198933.3 NP_945151.2 4 347
NM_198934.3 NP_945152.2 4 347
GenBank mRNA AF508939.1 - 4 256
AJ296212.1 - 4 299
AJ489474.1 - 4 347
AJ489475.1 - 4 338
AK028237.1 - 4 277
AY177625.1 - 4 256
BC066201.1 - 4 251
S65461.1 - 3 175
X68362.1 - 4 248
X68363.1 - 4 246
X68364.1 - 4 262
X70324.1 - 2 107

View Full Product Details