Gene Transcripts

Gene Symbol
Rbp1
Entrez Gene ID
19659
Gene Name
retinol binding protein 1, cellular
Gene Aliases
CRBPI, Crbp, Rbp-1
Chromosome Location
Chr. 9: 98422961 - 98446551 on Build GRCm38
UniGene ID
Mm.279741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011254.5 NP_035384.1 4 2453
GenBank mRNA AK011641.1 - 4 601
BC018254.1 - 4 580 3498107
X60367.1 - 4 2452

View Full Product Details