Gene Transcripts
- Gene Symbol
- CHRNE
- Entrez Gene ID
- 1145
- Gene Name
- cholinergic receptor nicotinic epsilon subunit
- Gene Aliases
- ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
- Chromosome Location
- Chr. 17: 4897769 - 4905019 on Build GRCh38
- UniGene ID
- Hs.654535
- Species
- Homo sapiens
- Gene Symbol
- C17orf107
- Entrez Gene ID
- 100130311
- Gene Name
- chromosome 17 open reading frame 107
- Gene Aliases
- Chromosome Location
- Chr. 17: 4899653 - 4902932 on Build GRCh38
- UniGene ID
- Hs.579243
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AW014012.1 |
- |
1 |
217 |
|
View Full Product Details