Gene Transcripts

Gene Symbol
Znrf4
Entrez Gene ID
20834
Gene Name
zinc and ring finger 4
Gene Aliases
1700094M01Rik, Ssrzf1, spzn
Chromosome Location
Chr. 17: 56511248 - 56512483 on Build GRCm38
UniGene ID
Mm.76862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011483.2 NP_035613.2 1 273
GenBank mRNA AB016984.1 - 1 207
AK005843.1 - 1 222
BC140986.1 - 1 156 9055598
BY104804.1 - 1 273

View Full Product Details