Gene Transcripts

Gene Symbol
Stat4
Entrez Gene ID
20849
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
Chromosome Location
Chr. 1: 51987106 - 52107189 on Build GRCm38
UniGene ID
Mm.1550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK037598.1 - 5 505
AK132675.1 - 7 692
AK157083.1 - 6 566
BC052907.1 - 6 571
BC098499.1 - 6 559 30074608
BY750146.1 - 6 557
U06923.1 - 6 562
U09351.1 - 6 1016

View Full Product Details