Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011595.2 | NP_035725.1 | 4 | 796 | |
GenBank mRNA | AB041611.2 | - | 4 | 653 | |
AK083905.1 | - | 4 | 654 | ||
AK128943.1 | - | 4 | 655 | ||
AK147847.1 | - | 4 | 654 | ||
BC014713.1 | - | 4 | 618 | 4239015 | |
L19622.1 | - | 4 | 395 | ||
L27424.1 | - | 4 | 665 | ||
Z30970.1 | - | 4 | 670 |