Gene Transcripts

Gene Symbol
Ikzf4
Entrez Gene ID
22781
Gene Name
IKAROS family zinc finger 4
Gene Aliases
A630026H08Rik, Eos, Zfpn1a4, Znfn1a4
Chromosome Location
Chr. 10: 128630839 - 128669358 on Build GRCm38
UniGene ID
Mm.358648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011772.2 NP_035902.2 4 560
GenBank mRNA AB017615.1 - 3 967
AK089522.1 - 4 394
AK165740.1 - 824
AK170453.1 - 5 484
BC141286.1 - 3 658 9055898
BC145614.1 - 3 644

View Full Product Details