Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011824.4 | NP_035954.1 | 2 | 281 | |
GenBank mRNA | AF045801.1 | - | 1 | 122 | |
AF108189.1 | - | 1 | 191 | ||
AK135022.1 | - | 2 | 316 | ||
AK157793.1 | - | 2 | 282 | ||
AK164871.1 | - | 2 | 282 | ||
BC015293.1 | - | 2 | 251 | 4216669 | |
BY151119.1 | - | 2 | 279 |