Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011891.4 | NP_036021.1 | 2 | 146 | |
GenBank mRNA | AB024923.1 | - | 2 | 367 | |
AK040974.1 | - | 2 | 148 | ||
AK047384.1 | - | 990 | |||
AK048480.1 | - | 3 | 331 | ||
BC130019.1 | - | 2 | 136 | 40140624 |