Gene Transcripts

Gene Symbol
Usp18
Entrez Gene ID
24110
Gene Name
ubiquitin specific peptidase 18
Gene Aliases
1110058H21Rik, AW047653, UBP43, Ubp15
Chromosome Location
Chr. 6: 121245906 - 121270917 on Build GRCm38
UniGene ID
Mm.326911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011909.2 NP_036039.2 2 350
GenBank mRNA AF069502.1 - 2 311
AK004296.1 - 2 320
AK149772.1 - 2 351
AK151011.1 - 2 351
AK151260.1 - 2 351
AK151722.1 - 2 351
AK152034.1 - 2 316
AK152574.1 - 2 316
AK153312.1 - 2 320
BC138562.1 - 2 232 8861582
BC138577.1 - 2 232 8861597
CT010228.1 - 1 86

View Full Product Details