Gene Transcripts
- Gene Symbol
- Phtf1
- Entrez Gene ID
- 18685
- Gene Name
- putative homeodomain transcription factor 1
- Gene Aliases
- AU041898, AW049785, Phft, Phtf
- Chromosome Location
- Chr. 3: 103968110 - 104007490 on Build GRCm38
- UniGene ID
- Mm.385178
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163467.1 |
NP_001156939.1
|
3, 4 |
265 |
|
NM_001163468.1 |
NP_001156940.1
|
3, 4 |
671 |
|
NM_001163469.1 |
NP_001156941.1
|
3, 4 |
671 |
|
NM_013629.2 |
NP_038657.2
|
3, 4 |
671 |
|
GenBank mRNA |
AJ133721.1 |
- |
3, 4 |
265 |
|
AJ242864.1 |
- |
3, 4 |
671 |
|
AK019146.1 |
- |
3, 4 |
575 |
|
AK088714.1 |
- |
3, 4 |
562 |
|
AK131629.1 |
- |
|
563 |
|
BC021147.1 |
- |
3, 4 |
543 |
|
BC050825.1 |
- |
3, 4 |
483 |
6518647 |
BC057340.1 |
- |
3, 4 |
557 |
6827816 |
BC060177.1 |
- |
3, 4 |
557 |
6827816 |
View Full Product Details