Gene Transcripts
- Gene Symbol
- Mkln1
- Entrez Gene ID
- 27418
- Gene Name
- muskelin 1, intracellular mediator containing kelch motifs
- Gene Aliases
- A130067F06Rik, AU015903, BB154892
- Chromosome Location
- Chr. 6: 31398828 - 31511795 on Build GRCm38
- UniGene ID
- Mm.238038
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013791.2 |
NP_038819.1
|
18 |
3707 |
|
GenBank mRNA |
U72194.1 |
- |
18 |
3710 |
|
View Full Product Details