Gene Transcripts

Gene Symbol
Rlim
Entrez Gene ID
19820
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
AL022832, AW743871, Ha1r, Rlim1, Rnf12
Chromosome Location
Chr. X: 103957164 - 103981297 on Build GRCm38
UniGene ID
Mm.427762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011276.3 NP_035406.3 5 5503
GenBank mRNA AF197105.1 - 1 1623
AK052543.1 - 1 1783
BC053711.1 - 1 1515
CB237411.1 - 1 276

View Full Product Details