Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015774.3 | NP_056589.1 | 2, 3 | 324 | |
GenBank mRNA | AK142595.1 | - | 2, 3 | 315 | |
AK159217.1 | - | 2, 3 | 291 | ||
BC025102.1 | - | 2, 3 | 276 | 2654098 | |
CT010195.1 | - | 2, 3 | 242 | ||
CT010259.1 | - | 2, 3 | 242 |