Gene Transcripts

Gene Symbol
Hpcal1
Entrez Gene ID
53602
Gene Name
hippocalcin-like 1
Gene Aliases
NVP-3, Nvp3, VILIP3, Vnsl3, Vsnl3
Chromosome Location
Chr. 12: 17690750 - 17791944 on Build GRCm38
UniGene ID
Mm.313619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF085192.1 - 4 1302
AK187818.1 - 1 53
AK210795.1 - 1 53
BC001997.1 - 4 1275 3496729

View Full Product Details