Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005267.4 | NP_005258.2 | 2 | 507 | |
XM_011509416.1 | XP_011507718.1 | 1 | 1254 | ||
XM_011509417.1 | XP_011507719.1 | 1 | 1254 | ||
GenBank mRNA | CD674975.1 | - | 2 | 507 | |
DB476698.1 | - | 1 | 144 | ||
KJ534845.1 | - | 1 | 454 |