Gene Transcripts
- Gene Symbol
- Letm1
- Entrez Gene ID
- 56384
- Gene Name
- leucine zipper-EF-hand containing transmembrane protein 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 33739675 - 33782704 on Build GRCm38
- UniGene ID
- Mm.260538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019694.1 |
NP_062668.1
|
6, 7 |
1225 |
|
GenBank mRNA |
AB041582.1 |
- |
6, 7 |
1238 |
|
AF061026.1 |
- |
6, 7 |
1225 |
|
AK133849.1 |
- |
6, 7 |
1244 |
|
AK206007.1 |
- |
1, 2 |
220 |
|
AK208931.1 |
- |
1, 2 |
201 |
|
AK213040.1 |
- |
1, 2 |
197 |
|
BC023862.1 |
- |
6, 7 |
1222 |
|
BC046326.1 |
- |
6, 7 |
1232 |
|
BC055865.1 |
- |
6, 7 |
1243 |
|
BC061115.1 |
- |
6, 7 |
1238 |
6533034 |
BC061690.1 |
- |
6, 7 |
1225 |
|
View Full Product Details