Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019821.2 | NP_062795.2 | 2 | 220 | |
GenBank mRNA | AF209703.1 | - | 2 | 146 | |
AK157313.1 | - | 2 | 207 | ||
BC016584.1 | - | 2 | 220 | 3157800 | |
BC053729.1 | - | 2 | 163 | 30065192 |