Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080383.1 | NP_001073852.1 | 3 | 728 | |
NM_005497.3 | NP_005488.2 | 3 | 732 | ||
XM_005256920.1 | XP_005256977.1 | 3 | 726 | ||
XM_005256921.1 | XP_005256978.1 | 2 | 554 | ||
GenBank mRNA | AK124339.1 | - | 2 | 966 | |
BC096213.3 | - | 1 | 458 | 40004800 | |
BC096214.3 | - | 1 | 458 | 40004801 | |
BC096215.3 | - | 1 | 458 | 40004803 | |
BC096216.3 | - | 1 | 458 | 40004809 | |
CR933719.1 | - | 1 | 50 |