Gene Transcripts

Gene Symbol
PLLP
Entrez Gene ID
51090
Gene Name
plasmolipin
Gene Aliases
PMLP, TM4SF11
Chromosome Location
Chr. 16: 57256097 - 57284672 on Build GRCh38
UniGene ID
Hs.632215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015993.2 NP_057077.1 4 1309
GenBank mRNA AF137386.1 - 4 1305
AK130584.1 - 4 1301
BC002760.2 - 4 1272 3633345

View Full Product Details