Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020578.3 | NP_065603.2 | 4 | 1025 | |
GenBank mRNA | AF155883.1 | - | 4 | 996 | |
AF506002.1 | - | 4 | 820 | ||
AK128964.1 | - | 4 | 1004 | ||
AK141265.1 | - | 4 | 1027 | ||
AK218582.1 | - | 2 | 137 | ||
BC046596.1 | - | 4 | 769 | 6312263 |