Gene Transcripts

Gene Symbol
Ap4s1
Entrez Gene ID
11782
Gene Name
adaptor-related protein complex AP-4, sigma 1
Gene Aliases
AI314282
Chromosome Location
Chr. 12: 51689888 - 51738939 on Build GRCm38
UniGene ID
Mm.386749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021710.3 NP_068356.1 6 876
GenBank mRNA AF092093.1 - 6 805
AK005283.1 - 6 626
BC053339.1 - 6 636 5370721
BP426557.1 - 4 331

View Full Product Details