Gene Transcripts
- Gene Symbol
- Rhot1
- Entrez Gene ID
- 59040
- Gene Name
- ras homolog gene family, member T1
- Gene Aliases
- 2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1
- Chromosome Location
- Chr. 11: 80209005 - 80267907 on Build GRCm38
- UniGene ID
- Mm.489674
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163354.1 |
NP_001156826.1
|
5, 6 |
516 |
|
NM_001163355.1 |
NP_001156827.1
|
5, 6 |
516 |
|
NM_021536.7 |
NP_067511.4
|
5, 6 |
516 |
|
GenBank mRNA |
AK019059.1 |
- |
5, 6 |
476 |
|
AK028597.1 |
- |
5, 6 |
459 |
|
AK031139.1 |
- |
3, 4 |
178 |
|
AK032096.1 |
- |
5, 6 |
489 |
|
AK041119.1 |
- |
5, 6 |
452 |
|
AK054472.1 |
- |
5, 6 |
487 |
|
AK078955.1 |
- |
5, 6 |
459 |
|
AK138830.1 |
- |
5, 6 |
457 |
|
AK141621.1 |
- |
5, 6 |
343 |
|
AK147749.1 |
- |
5, 6 |
495 |
|
AK147801.1 |
- |
5, 6 |
521 |
|
AK167502.1 |
- |
5, 6 |
516 |
|
AK169811.1 |
- |
5, 6 |
485 |
|
AK191854.1 |
- |
3, 4 |
150 |
|
BC046785.1 |
- |
5, 6 |
499 |
6416127 |
BC058350.1 |
- |
5, 6 |
431 |
6815832 |
View Full Product Details