Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042659.1 | NP_001036124.1 | 2 | 1375 | |
NM_022721.3 | NP_073558.2 | 2 | 1482 | ||
GenBank mRNA | AB052910.1 | - | 1 | 608 | |
AF272146.1 | - | 1 | 599 | ||
AK030111.1 | - | 2 | 1484 | ||
AK039402.1 | - | 2 | 1375 | ||
BC117723.2 | - | 2 | 908 | 40088671 |