Gene Transcripts

Gene Symbol
Txnip
Entrez Gene ID
56338
Gene Name
thioredoxin interacting protein
Gene Aliases
1200008J08Rik, AA682105, Hyplip1, THIF, Tbp-2, VDUP1
Chromosome Location
Chr. 3: 96557957 - 96561857 on Build GRCm38
UniGene ID
Mm.410189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009935.2 NP_001009935.1 8 1489
NM_023719.2 NP_076208.2 8 1486
GenBank mRNA AF173681.1 - 8 1250
AK004653.2 - 8 1486
AK082476.1 - 8 1484
AK088870.1 - 8 1484
AK089304.1 - 8 1489
AK089351.1 - 8 1487
AK089403.1 - 8 1489
AK155517.1 - 8 1487
AK171205.1 - 8 1487
AK171312.1 - 8 1487
AK171425.1 - 8 1487
AK171456.1 - 8 1484
AK171464.1 - 8 1487
AK171527.1 - 8 1487
BC011212.1 - 8 1474 4193517
BC031850.1 - 8 1467 3591421

View Full Product Details