Gene Transcripts

Gene Symbol
Rab5c
Entrez Gene ID
19345
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
AI326010, Rabl
Chromosome Location
Chr. 11: 100715009 - 100738210 on Build GRCm38
UniGene ID
Mm.29829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK089199.1 - 6 1926
AK134691.1 - 6 1812
AK167599.1 - 6 1861

View Full Product Details