Gene Transcripts
- Gene Symbol
- Ublcp1
- Entrez Gene ID
- 79560
- Gene Name
- ubiquitin-like domain containing CTD phosphatase 1
- Gene Aliases
- 4930527B16Rik, 8430435I17Rik, BC002236
- Chromosome Location
- Chr. 11: 44454571 - 44470548 on Build GRCm38
- UniGene ID
- Mm.338784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024475.5 |
NP_077795.2
|
10, 11 |
1101 |
|
GenBank mRNA |
AK043952.1 |
- |
10, 11 |
1008 |
|
AK050031.1 |
- |
10, 11 |
1012 |
|
AK147744.1 |
- |
10, 11 |
1049 |
|
AK153998.1 |
- |
10, 11 |
1013 |
|
BC002236.1 |
- |
3, 4 |
375 |
|
BC056652.1 |
- |
6, 7 |
602 |
|
BC083331.1 |
- |
8, 9 |
762 |
|
BC085111.1 |
- |
9, 10 |
1004 |
30735741 |
BC106093.1 |
- |
9, 10 |
978 |
5098953 |
View Full Product Details