Gene Transcripts

Gene Symbol
Exoc2
Entrez Gene ID
66482
Gene Name
exocyst complex component 2
Gene Aliases
2410030I24Rik, AI648199, Sec5, Sec5l1
Chromosome Location
Chr. 13: 30789516 - 30974273 on Build GRCm38
UniGene ID
Mm.293510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025588.2 NP_079864.1 3 317
GenBank mRNA AK016532.1 - 4 444
AK032766.1 - 3 339
AK042744.1 - 3 364
AK050068.1 - 3 325
AK180355.1 - 2 210
BC048154.1 - 3 317 6400932
BC049102.1 - 3 317 5702581

View Full Product Details