Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000073.2 | NP_000064.1 | 3 | 184 | |
XM_005271724.3 | XP_005271781.1 | 3 | 221 | ||
XM_006718941.2 | XP_006719004.1 | 3 | 172 | ||
GenBank mRNA | AK223314.1 | - | 3 | 137 | |
AK299997.1 | - | 2 | 876 | ||
AK310578.1 | - | 4 | 312 | ||
AK310622.1 | - | 4 | 342 | ||
AK313966.1 | - | 3 | 172 | ||
AK315895.1 | - | 3 | 128 | ||
BC113830.1 | - | 3 | 130 | 40029587 | |
X04145.1 | - | 3 | 141 | ||
X60492.1 | - | 2, 3 | 124 |