Gene Transcripts

Gene Symbol
Tmem41a
Entrez Gene ID
66664
Gene Name
transmembrane protein 41a
Gene Aliases
2900010K02Rik, 5730578N08Rik
Chromosome Location
Chr. 16: 21934327 - 21947552 on Build GRCm38
UniGene ID
Mm.46480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025693.4 NP_079969.1 5 860
GenBank mRNA AK007685.1 - 5 855
AK013518.1 - 4 685
AK160292.1 - 5 781
BC019770.1 - 5 815 5124769

View Full Product Details