Gene Transcripts

Gene Symbol
Ttc39b
Entrez Gene ID
69863
Gene Name
tetratricopeptide repeat domain 39B
Gene Aliases
1810054D07Rik, 9130422G05Rik, AI450603, AV103077, AW215585
Chromosome Location
Chr. 4: 83220301 - 83324270 on Build GRCm38
UniGene ID
Mm.5540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027238.2 NP_081514.1 20 6898
GenBank mRNA AK021142.1 - 1 1035
AK031930.1 - 1 2961
AK134680.1 - 1 869
BC151021.1 - 20 6871 9087934


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_025782.1 - 2961 Chr. - - [-]

View Full Product Details