Gene Transcripts

Gene Symbol
Lsm14a
Entrez Gene ID
67070
Gene Name
LSM14 homolog A (SCD6, S. cerevisiae)
Gene Aliases
2700023B17Rik, AA407828, AU017544, Tral
Chromosome Location
Chr. 7: 34344637 - 34389637 on Build GRCm38
UniGene ID
Mm.276503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025948.2 NP_080224.1 5, 6 780
GenBank mRNA AK003611.1 - 3, 4 340
AK012276.1 - 4, 5 543
AK186175.1 - 2, 3 300
AK202607.1 - 1, 2 116
BC031521.1 - 5, 6 780 2655161

View Full Product Details