Gene Transcripts

Gene Symbol
Naaa
Entrez Gene ID
67111
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
2210023K21Rik, 3830414F09Rik, Asahl
Chromosome Location
Chr. 5: 92257660 - 92278181 on Build GRCm38
UniGene ID
Mm.28890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163687.1 NP_001157159.1 4 613
NM_025972.4 NP_080248.2 4 619
GenBank mRNA AB162194.1 - 4 585
AK008776.1 - 4 621
AK017141.1 - 4 598
AK133485.1 - 4 604
AK133658.1 - 4 621
AK146191.1 - 4 607
BC004572.1 - 4 589 3586255

View Full Product Details