Gene Transcripts

Gene Symbol
Pi4k2b
Entrez Gene ID
67073
Gene Name
phosphatidylinositol 4-kinase type 2 beta
Gene Aliases
2610042N09Rik, 4933409G22Rik
Chromosome Location
Chr. 5: 52741557 - 52769344 on Build GRCm38
UniGene ID
Mm.248647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025951.3 NP_080227.2 6, 7 1095
NM_028744.3 NP_083020.2 6, 7 1026
GenBank mRNA AF411321.1 - 6, 7 950
AK011751.1 - 6, 7 1058
AK016754.1 - 6, 7 989
AK035523.1 - 6, 7 1080
AK170730.1 - 6, 7 1079
AY148879.1 - 989
BC010257.1 - 103
BC062144.1 - 6, 7 1002 6335661

View Full Product Details