Gene Transcripts

Gene Symbol
Tomm34
Entrez Gene ID
67145
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
2610100K07Rik, TOM34
Chromosome Location
Chr. 2: 164053538 - 164071177 on Build GRCm38
UniGene ID
Mm.23173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291155.1 NP_001278084.1 2 327
NM_025996.5 NP_080272.1 2 706
GenBank mRNA AK011786.1 - 2 706
AK017699.1 - 2 272
AK075873.1 - 2 324
AK168329.1 - 2 261
BB869739.1 - 2 328
BC018278.1 - 2 257 4187356
CX235437.1 - 2 246

View Full Product Details