Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026092.3 | NP_080368.1 | 4, 5 | 513 | |
GenBank mRNA | AK006241.1 | - | 4, 5 | 514 | |
AK006630.1 | - | 4, 5 | 514 | ||
AY572447.1 | - | 4, 5 | 472 | ||
BC048450.1 | - | 4, 5 | 484 | 6774504 |