Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026220.4 | NP_080496.1 | 2 | 222 | |
GenBank mRNA | AK012688.1 | - | 2 | 208 | |
AK014483.1 | - | 2 | 215 | ||
AK032586.1 | - | 2 | 223 | ||
AK050561.1 | - | 2 | 223 | ||
AK077604.1 | - | 2 | 216 | ||
AK088766.1 | - | 2 | 219 | ||
AK161000.1 | - | 2 | 223 | ||
AK168801.1 | - | 2 | 228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081975.3 | NP_001075444.1 | 2 | 218 | |
GenBank mRNA | AK011317.1 | - | 2 | 216 | |
AK018514.1 | - | 2 | 219 | ||
AK088946.1 | - | 225 | |||
AK169029.1 | - | 226 | |||
BC005728.1 | - | 2 | 179 | 3595848 |