Gene Transcripts
- Gene Symbol
- Snrnp48
- Entrez Gene ID
- 67797
- Gene Name
- small nuclear ribonucleoprotein 48 (U11/U12)
- Gene Aliases
- 1110050F08Rik, 6530403A03Rik, C6orf151
- Chromosome Location
- Chr. 13: 38204911 - 38227663 on Build GRCm38
- UniGene ID
- Mm.250783
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026382.2 |
NP_080658.2
|
1, 2 |
198 |
|
GenBank mRNA |
AK007431.1 |
- |
1, 2 |
187 |
|
AK012916.1 |
- |
1, 2 |
193 |
|
AK018315.1 |
- |
1, 2 |
193 |
|
AK036713.1 |
- |
1, 2 |
198 |
|
AK087789.1 |
- |
1, 2 |
193 |
|
AK172155.1 |
- |
1, 2 |
187 |
|
BC021443.1 |
- |
1, 2 |
181 |
5038605 |
BC037073.1 |
- |
1, 2 |
188 |
|
BY263553.1 |
- |
1, 2 |
201 |
|
View Full Product Details