Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
67842
Gene Name
NOP9 nucleolar protein
Gene Aliases
2610027L16Rik
Chromosome Location
Chr. 14: 55745693 - 55755634 on Build GRCm38
UniGene ID
Mm.180338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026403.3 NP_080679.3 1 269
GenBank mRNA AK011570.1 - 1 259
AK032873.1 - 1 253
AK037831.1 - 1 257
AK050912.1 - 1 259
AK083861.1 - 1 138
AK139689.1 - 1 219
AK146020.1 - 1 224
AK146687.1 - 1 240
AK152732.1 - 1 243
AK154387.1 - 1 261
AK159798.1 - 1 243
BC018523.1 - 1 249 3499986
CJ098414.1 - 1 270

View Full Product Details